missing translation for 'onlineSavingsMsg'
Learn More

BOLA Miniature Screw-in Adaptors

Product Code. 10053241
Change view
Click to view available options
Connections:
Female
Male
Diameter (Metric) Inner:
0.8 mm
1.6 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections Diameter (Metric) Inner unitSize
10053241 Male 0.8 mm Each
10459191 Female 0.8 mm Each
10574972 Male 1.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053241 Supplier BOLA Supplier No. F71606
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Miniature Screw-in Adaptors PTFE, UNF 1/4 28G female, NPT 1/4 in.

Adaptor made of PTFE for transition from female thread UNF ¼ in. 28 G to male thread NPT 1/8 in. or NPT ¼ in..

Specifications

Diameter (Metric) Outer 1.6 mm
Connections Male
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.