Learn More
BOLA Miniature Manifold Blocks
Product Code. 10165061
Change view
Click to view available options
Diameter (Metric) Inner:
0.8 mm
Unit Size:
Each
Product Code. Diameter (Metric) Inner unitSize
10165061 0.8 mm Each
1 options
Product Code. 10165061 Supplier BOLA Supplier No. F71210
Only null left
Add to basket
Add to basket
Miniature Manifold Blocks PTFE, 4 stopcocks, ports:1 inlet, 4 outlets
  • Block made of PTFE with one inlet and four outlets with female thread UNF ¼ in. 28 G.
  • Also available with four stopcocks with stopcock plugs made of FEP for easy interruption and control of flow.
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 0.8 mm
Diameter (Metric) Outer 1.6 mm