missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA Miniature Couplings
Description
- Three types of couplings available: straight type made of PTFE or POM with higher mechanical strength
- T type with three connections or cross type with four connections, all made of PTFE.
- The tubing is connected by means of tube end fittings to a female thread UNF ¼ in. 28 G.
Specifications
Specifications
| Diameter (Metric) Outer | 1.6 mm |
| Connections | 3 |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.8 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?