missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Miltex™ Half-curved Iris Tissue Forceps

Product Code. 11770309 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (Metric):
10.16 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11770309 10.16 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11770309 Supplier Cole-Parmer™ Supplier No. 1092304

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Feature the highest quality stainless steel, crafted to the most exacting standards

  • Precision cutting and holding surfaces
  • Ratcheting lock mechanisms
  • Non-glare finish

Specifications

Tip Style Curved
Length (English) 4 in.
Material Stainless Steel
Length (Metric) 10.16 cm
Product Type Precision-grade Iris Tissue Forceps
Grade Precision-grade
Type Iris Tissue Forceps
Description Iris Tissue Forceps, Half curved, Precision-grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.