missing translation for 'onlineSavingsMsg'
Learn More

Kartell SpA Micropipette Tips

Product Code. 10214222
Change view
Click to view available options
Format:
Bagged
Rack
Unit Size:
Pack of 200
Pack of 2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
10214222 Bagged Pack of 2000
10172362 Rack Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214222 Supplier Kartell SpA Supplier No. 0094200
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Micropipette tips, 500 to 5000 μL

Manufactured from virgin cadmium-free polypropylene. High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum. The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered.

Recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market. Compatibility: Finnpipette™, Brand™

Specifications

Volume (Metric) 500 to 5000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Translucent
Autoclavable Autoclavable
Format Bagged
Tip Style Low Retention
Product Type Tips
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line Dispolab
Material Virgin Cadmium-free Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.