Learn More
Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates
GREENER_CHOICE
Product Code. 10033281
Change view
Click to view available options
Binding Property:
High Binding
Medium Binding
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Binding Property unitSize
10033281 Medium Binding Pack of 100
10063291 High Binding Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10033281 Supplier Greiner Bio-One Supplier No. 701070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use in flexibility in diagnostics. Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates are suitable for transmission measurements.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • For optical measurements
  • Measuring light beam is not deflected by well profile
  • Specifically treated to provide incresed protein binding
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.