missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies Micro Flow cell 2 uL, 3mm path, 120 bar

Brand:  Agilent Technologies G1315-60024

Encompass_Preferred

Micro Flow cell 2 uL, 3mm path, 120 bar

Product Code. 10032323

  • £3052.35 / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.