Learn More
Mettler Toledo™ Antistatic Kit for XP Analytical Balances
Product Code. 12704592 Shop All Mettler Toledo Products
Change view
Click to view available options
:
Antistatic Kit; For Mettler XP Analytical Balances
Product Code.
12704592 Antistatic Kit; For Mettler XP Analytical Balances
1 options
Product Code. 12704592
Brand: Mettler Toledo™ 11107761
For use with XP Analytical Balances.
For discharging electrostatically charged weighing containers and weighing objects (single point ionizer).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XP Analytical Balance
Includes Point electrode, XP power supply
Type Anti-static Kit