missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfide, 99+%, extra pure

Product Code. 10071460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071460 1 L Glass bottle 1 L
10071470 10 mL Glass bottle 10mL
10264790 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071460 Supplier Thermo Scientific Acros Supplier No. 127760010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-18-3 | C2H6S | 62.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
IUPAC Name methylsulfanylmethane
SMILES CSC

Specifications

Melting Point -98.0°C
Color Colorless
Density 0.8460g/mL
Boiling Point 38.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. Methylmercaptan (GC)
Packaging Glass bottle
Linear Formula (CH3)2S
Refractive Index 1.4351
Quantity 1 L
Beilstein 01, 288
Fieser 02,156; 04,190; 05,260; 15,88; 16,84; 17,198
Merck Index 15, 3281
Specific Gravity 0.846
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 62.13
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methyl sulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl sulfide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.