missing translation for 'onlineSavingsMsg'
Learn More

Methyl (+)-4-Boc-piperazine-2-carboxylate

Product Code. 11397509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397509 1 g 1g
11307519 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397509 Supplier Thermo Scientific Alfa Aesar Supplier No. H27886.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 129799-08-2 | C11H21N2O4 | 245.30 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129799-08-2
Molecular Formula C11H21N2O4
Molecular Weight (g/mol) 245.30
MDL Number MFCD01632465,MFCD04115325,MFCD04115326
InChI Key QUKAHFCVKNRRBU-QMMMGPOBSA-O
Synonym methyl 4-boc-piperazine-2-carboxylate, 1-tert-butyl 3-methyl piperazine-1,3-dicarboxylate, n-4-boc-2-piperazinecarboxylic acid methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester, 4n-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, 1,3-piperazinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester; piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, tert-butyl 3-methoxycarbonyl piperazinecarboxylate, pubchem18338
PubChem CID 2756819
IUPAC Name 1-O-tert-butyl 3-O-methyl piperazine-1,3-dicarboxylate
SMILES COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 244.29
Chemical Name or Material Methyl (+/-)-4-Boc-piperazine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.