missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetate Hydrogen Sulfate ((R)-Clopidogrel Hydrogen Sulfate), Mikromol™

Product Code. 17972726
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972726 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17972726 Supplier Mikromol Supplier No. MM0455.120025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetate Hydrogen Sulfate ((R)-Clopidogrel Hydrogen Sulfate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Clopidogrel Hydrogen Sulfate
CAS 120202-71-3
Analyte or Component Names (R)-Clopidogrel Hydrogen Sulfate
Shipping Condition Room Temperature
Molecular Formula C16 H16 Cl N O2 S . H2 O4 S
Certifications/Compliance ISO 17025
Synonym Methyl (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetate Hydrogen Sulfate, Clopidogrel Hydrogen Sulfate Imp. C (EP), Clopidogrel Imp. C (EP), Clopidogrel USP Related Compound C, Clopidogrel USP RC C, Clopidogrel Hydrogen Sulfate Imp. C (EP) as Hydrogen Sulfate, (R)-Clopidogrel Hydrogen Sulfate, Clopidogrel Related Compound C as Sulfate, Clopidogrel Hydrogen Sulfate Impurity C as Hydrogen Sulfate, Clopidogrel Impurity C as Hydrogen Sulfate
InChI Formula InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m1./s1
SMILES COC(=O)[C@H](N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O
IUPAC Name methyl (2R)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
Molecular Weight (g/mol) 419.9
Quantity 25 mg
Formula Weight 419.0264 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.