missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate, Mikromol

Product Code. 30463766
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463766 4 x 25 mg -
17960756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30463766 Supplier Mikromol Supplier No. MM3741.0200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate
CAS 121659-86-7
Analyte or Component Names Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate
Shipping Condition Room Temperature
Molecular Formula C20 H16 F N O2
Certifications/Compliance ISO 17025
Synonym 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylic Acid Methyl Ester, 2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylic acid methyl ester, 4-(4-Fluorophenyl)-2-cyclopropylquinoline-3-carboxylic acid methyl ester, Methyl 2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate
InChI Formula InChI=1S/C20H16FNO2/c1-24-20(23)18-17(12-8-10-14(21)11-9-12)15-4-2-3-5-16(15)22-19(18)13-6-7-13/h2-5,8-11,13H,6-7H2,1H3
Solution Type Neat
SMILES COC(=O)c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4
IUPAC Name methyl 2-cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylate
Molecular Weight (g/mol) 321.34
Quantity 4 x 25 mg
Formula Weight 321.1165 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.