missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10179100 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10179100 2.5 L Glass Bottle 2.5 L
10281001 1 L Glass Bottle 1 L
15538464 2.5 L Plastic Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10179100 Supplier JT Baker Supplier No. 8045.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Max. 10 (APHA)
Appearance Clear
Density 0.7918 g/cm³ (20°C)
Boiling Point 64.6°C (1013 hPa)
Quantity 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Assay Percent Range min. 99.8 % (by GC, corrected for water)
Residue after Evaporation max. 0.001 %
Water max. 0.05 %
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.