missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) nitrate monohydrate, 98+%

Product Code. 10195733 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195733 1 kg Plastic bottle 1kg
10145080 50 g Plastic bottle 50g
10115130 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code. 10195733 Supplier Thermo Scientific Acros Supplier No. 213140010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-34-8
Molecular Formula H2HgN2O7
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149736
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
Synonym mercuric nitrate monohydrate, unii-yj09edi5uy, mercury ii nitrate hydrate, yj09edi5uy, mercury 2+ ion hydrate dinitrate, nitric acid, mercury 2+ salt, monohydrate, mercury nitrate monohydrate, 324.60 anhydrous, mercury 2+ dinitrate hydrate, hgn2o6.xh2o x=1-2
PubChem CID 3084029
IUPAC Name mercury(2+) hydrate dinitrate
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 77.0°C to 79.0°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 1 kg
Assay Percent Range 98% min. (Complexometry)
Linear Formula Hg(NO3)2·H2O
Fieser 03,197; 10,254; 11,317
Merck Index 14, 5880
Solubility Information Solubility in water: slightly soluble
Formula Weight 342.6
Percent Purity ≥98%
Packaging Plastic bottle
Specific Gravity 4.3
Density 4.3000g/mL
Chemical Name or Material Mercury(II) nitrate monohydrate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(II) nitrate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.