missing translation for 'onlineSavingsMsg'
Learn More

Mepiquat Iodide D3 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321266
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321266 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321266 Supplier Dr. Ehrenstorfer Supplier No. DREA14880100AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mepiquat methyl-d3 iodide
CAS 32317-85-4
Solvent or Matrix Acetonitrile
Molecular Formula C7 2H3 H13 N . I
Quantity 1 mL
Synonym 1-Methyl-1-methyl-d3-piperidinium Iodide, Mepiquat iodide D3 (methyl-d3)
InChI Formula InChI=1S/C7H16N.HI/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1/i1D3;
SMILES [I-].[2H]C([2H])([2H])[N+]1(C)CCCCC1
IUPAC Name 1-methyl-1-(trideuteriomethyl)piperidin-1-ium;iodide
Molecular Weight (g/mol) 244.13
Formula Weight 244.0516 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.