missing translation for 'onlineSavingsMsg'
Learn More

Melaminsulfone Sodium Salt, TRC

Product Code. 30132819
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132819 2.5 g 2.5g
17973015 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132819 Supplier Toronto Research Chemicals Supplier No. TRCM2151152.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-89-5
Molecular Formula C12 H14 N3 O4 S . Na
Molecular Weight (g/mol) 319.31
Synonym [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt, 4-N-Desmethylmetamizole Sodium Salt, Melaminsulfone Sodium Salt, Metamizol Sodium Imp. E (EP) as Sodium Salt
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)amino]methanesulfonate
SMILES [Na+].CN1N(C(=O)C(=C1C)NCS(=O)(=O)[O-])c2ccccc2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H15N3O4S.Na/c1-9-11(13-8-20(17,18)19)12(16)15(14(9)2)10-6-4-3-5-7-10;/h3-7,13H,8H2,1-2H3,(H,17,18,19);/q;+1/p-1
Formula Weight 319.0603
Percent Purity >95%
Chemical Name or Material ((1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino)methanesulfonic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.