missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Medium 199, Hanks' Balanced Salts, powder
Medium 199, Hanks' Balanced Salts, powder
Brand: Gibco™ 10012011
Code : 50
Additional Details : Weight : 0.05250kg
Description
Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. This Medium 199 formulation contains Hanks' salts for use without CO2.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing
Gibco™ Medium 199 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Medium 199 contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This Medium 199 formulation does not require a CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Specifications
ISO 13485 standards, FDA | |
Hanks' Balanced Salts, Medium 199, 1L, Powdered Form, L-Glutamine, HEPES Buffer, 2 to 8°C Storage Temperature, 36 Months Shelf Life, For Mammalian Cell Culture, Vaccine Production and In-vitro Cultivation | |
Medium 199 Hanks' Balanced Salts | |
No Sodium Bicarbonate | |
7 to 7.4 |
2 to 8°C | |
Powder | |
Hanks’ Balanced Salt solution (HBSS), Glutamine, HEPES, Phenol Red | |
For cell culture, mammalian cell culture, vaccine production and In-vitro cultivation | |
1 L |