Learn More
WTW™ Measuring Vessel for Electrodes
GREENER_CHOICE
Product Code. 10079630
Change view
Click to view available options
For Use With (Application):
Determining pH of drinking water
Unit Size:
Each
Product Code. For Use With (Application) unitSize
10079630 Determining pH of drinking water Each
1 options
Product Code. 10079630 Supplier WTW™ Supplier No. 108062
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For determining the pH of drinking water.
  • Suitable for pH combination electrodes SenTix™ 81
  • Also for use as a storage vessel for electrodes
  • Supplied individually
TRUSTED_SUSTAINABILITY
Specifications
Type Measuring Vessel
For Use With (Application) Determining pH of drinking water