Learn More
Lovibond™ MD 100 Single-Parameter Photometer: High Range Manganese
Description
Includes
Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Vario set (manganese citrate buffer F10 and sodium periodate F10 powder, 100 tests), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
The calibration and software-based adjustment options mean that the MD 100 is also suitable for use as a testing instrument. The tests are conducted using either Lovibond tablet reagents with long term stability and a guaranteed minimum five or 10 year shelf life, VARIO powder reagents or using liquid reagents.
Features
- Waterproof (as defined in IP68, 1 hour at 0.1m)
- Automatic switch-off
- Real-time clock and date calibration mode
- Backlit LCD display (on keypress)
- Factory calibration and user calibration; Reset to factory calibration possible
- Internal ring memory for 16 data sets
- Infrared interface for test data transfer
Specifications
- Range: 0.1 to 18mg/L Mn
- Light source: LED, interference filter (IF) and photosensor in transparent cell chamber
- Wavelength Accuracy: ±1nm
- Photometric Accuracy: 3% FS (T = 20°C to 25°C)
- Photometric Resolution: 0.01 A
- Temperature: 5° to 40°C
- Relative Humidity: 30 to 90% (non condensing)
- Power Supply: 4 AAA batteries
- Operating Time: approx. 17 hours or 5000 test measurements in continuous mode when display backlight is off
- Dimensions (L x W x H): 15.5 x 7.5 x 3.5cm (6.1 x 3 x 1.5 in.)
- Weight: basic unit approx. 260g
Note
Specifications for each type of tablet as well as a Certificate of Analysis for each lot is available in the download area at www.lovibond.com.
Specifications
Specifications
| Type | MD 100 Photometer: High Range Manganese |
| Display Type | Backlit LCD |
| Accuracy | 3% FS |
| Height (Metric) | 3.5 cm |
| Includes | Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Vario set (manganese citrate buffer F10 and sodium periodate F10 powder, 100 tests), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual |
| Length (Metric) | 15.5 cm |
| Memory | Internal ring memory for 16 data sets |
| Light Source | LED |
| Range | 0.1 to 18mg/L Mn |
| Resolution | 0.01 A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.