missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MCF-7 Whole Cell Lysate

Product Code. 18093549 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093549 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093549 Supplier Novus Biologicals™ Supplier No. NBP2039640.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MCF7 (Human Breast Adenocarcinoma) whole breast cell lysate 2mg/mL supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. Growth media: ATCC medium. Lysates are guaranteed for 6 months from date of receipt.

The MCF-7 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue MCF-7
Quantity 0.1 mg
Storage Buffer Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol.
Test Specificity MCF7 (Human Breast Adenocarcinoma)
Host Species Human
Concentration 2 mg/mL
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.