missing translation for 'onlineSavingsMsg'
Learn More

MCB-613, MedChemExpress

Product Code. 16473777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473777 50 mg 50mg
16470288 5 mg 5mg
16443666 25 mg 25mg
16427828 10 mg 10mg
16417656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473777 Supplier Medchem Express Supplier No. HY19625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MCB-613 is a potent Steroid receptor coactivator SRC small molecule ‘stimulator’ (SMS), super-stimulates SRCs’ transcriptional activity. MCB-613 increases SRCs’ interactions with other coactivators and markedly induces ER stress coupled to the generation of reactive oxygen species (ROS). MCB-613 is a SMS that target oncogenes can be exploited as anti-cancer agents by over-stimulating the SRC oncogenic program.

MCB-613 is a potent Steroid receptor coactivator SRC small molecule ‘stimulator’ (SMS), super-stimulates SRCs’ transcriptional activity.

Specifications

Chemical Name or Material MCB-613
Physical Form Solid
CAS 1162656-22-5
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20N2O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (164.26 mM; Need ultrasonic)
SMILES O=C1/C(CC(CC)C/C1=C\C2=CC=CN=C2)=C/C3=CC=CN=C3
Molecular Weight (g/mol) 304.39
Formula Weight 304.39
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.