Learn More
BOLA Maxi Propeller Blades
Product Code. 10080772
Change view
Click to view available options
Dimensions (L x W x H):
140 x 20 x 4 mm
200 x 20 x 6 mm
200 x 26 x 6 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10080772 140 x 20 x 4 mm Each
16229621 200 x 20 x 6 mm Each
10326152 200 x 26 x 6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10080772 Supplier BOLA Supplier No. C44110
Only null left
Add to basket
Add to basket
Maxi Propeller Blades for stainless steel shafts jacketed with PTFE, glass shafts or metal shafts
  • These solid stirrer blades are made of PTFE and have a set of clamp screws made of a PTFE/PEEK compound.
  • The blades can be fixed tightly on BOLA Stirrer Shafts by means of the clamp screws. A spanner wrench is included for easy assembly.
Specifications
Dimensions (L x W x H) 140 x 20 x 4 mm
Coating PTFE