Learn More
Greiner Bio-One MASTERBLOCK™ 384 Deep Well Polypropylene Conical Bottom Microplates
Product Code. 10000222
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
10000222 Sterile Pack of 60
10124531 Non-sterile Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10000222 Supplier Greiner Bio-One Supplier No. 781271
Only null left
Add to basket
Add to basket
Free of detectable DNase, RNase, human DNA.
  • Alphanumeric well coding
  • High chemical resistanceHigh temperature resistanceSealable with adhesive films and heat sealer
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Binding Type Non Binding
Color Clear
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 20 to 225 μL
Sterility Sterile
Total Volume (Metric) Well 240 μL
Well Shape Rounded Square, Conical Bottom (V-well)
No. per Pack 6
Well Color Clear