Learn More
Invitrogen™ MARCO Monoclonal Antibody (PLK-1)
Description
Endotoxin: 0.40 EU/mg Application Notes: IHC (frozen) - tissue sections can be fixed in acetone or 2% paraformaldehyde. Western blot - use non-reducing conditions. Flow - this antibody weakly stains alveolar macrophages by recognizing the extracellular domain of MARCO, with transfected COS cells used as a positive control. Immunoprecipitation - will immunoprecipitate MARCO as a 60 kDa and 50 kDa protein from transfected COS lysates. Functional Assays - blocks human alveolar macrophages by recognizing the extracellular doamin of MARCO. Recommended positive control: Human alveolar macrophages; Negative control: All other human cells.
Specifications
Specifications
| Antigen | MARCO |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | PLK-1 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | Marco |
| Gene Accession No. | Q9UEW3 |
| Gene Alias | AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.