Learn More
Invitrogen™ Human MAPK8 (JNK1), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | P45983 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.05% Triton X-100, 0.05 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5599 |
| Molecular Weight (g/mol) | 47.6 kDa |
| Name | Human MAPK8 (JNK1), His Tag |
| Quantity | 1 mg |
| Regulatory Status | For research use only. Not for use in diagnostic procedures |
| Gene Alias | AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8 b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1 c; Stress-activated protein kinase JNK1; zgc:112379 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.