Learn More
Hamilton™ Manual HPLC Cemented Needle Syringes
Product Code. p-7138549
Change view
Click to view available options
Connection Type:
Cemented
Luer Tip, Cemented
Unit Size:
Each
Volume (Metric):
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type unitSize Volume (Metric)
11598044 Cemented Each -
11568054 Luer Tip, Cemented Each 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11598044 Supplier Hamilton™ Supplier No. DS80365
Specifically designed for manual or automatic injection into high pressure ports with Rheodyne™, Valco™, Beckman™ and SSI™ valves.
  • Needle is blunt and electro-polished to slide smoothly into the injection port
  • Available as standard cemented needle or Luer tip cemented needle
  • 51mm in length with point style 3
Specifications
Needle Gauge 22s G
Length (Metric) Needle 51 mm
Needle Point Style 3
Connection Type Cemented