missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Nitrate Tetrahydrate, ≥97.0% (KT), Honeywell™

Product Code. p-7120545 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15656090 100 g Glass Bottle 100g
15666090 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15656090 Supplier Honeywell Research Chemicals Supplier No. 63547100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥97.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20694-39-7
Molecular Formula H8MnN2O10
Molecular Weight (g/mol) 251.006
MDL Number MFCD00149789
InChI Key ALIMWUQMDCBYFM-UHFFFAOYSA-N
Synonym manganese ii nitrate tetrahydrate, manganese nitrate tetrahydrate, unii-03m0g68r5f, manganese 2+ ion tetrahydrate dinitrate, manganese dinitrate tetrahydrate, acmc-20akkk, nitric acid, manganese 2+ salt, tetrahydrate, ksc206g2f, mn.2no3.4h2o, manganese 2+ dinitrate tetrahydrate
PubChem CID 182616
IUPAC Name manganese(2+);dinitrate;tetrahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Mn+2]

Specifications

Color Pink
pH Acidic
Physical Form Solid
Quantity 100 g
Linear Formula Mn(NO3)2 · 4H2O
UN Number UN2724
Formula Weight 251.01g/mol
Percent Purity ≥97.0% (KT)
Odor Odorless
Packaging Glass Bottle
Chemical Name or Material Manganese(II) nitrate tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.