Learn More
Chem-Lab Mangan AAS-Standard, 4,569 g Mn(NO3)2x4H2Oin 2-5%, 500ml
Brand: Chem-Lab CL01.1316.0500
Mangan AAS-Standard, 4,569 g Mn(NO3)2x4H2Oin 2-5%, 500ml
Safety and Handling
- Manganese standard solution 1000 µg/ml
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Nitric acid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.