missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Malleable Cast Iron Micro Retort Clamp

Product Code. 10156410
Change view
Click to view available options
Diameter (Metric):
15 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10156410 15 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10156410 Supplier Bochem™ Supplier No. 5460
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1 Ea.
Type Micro Clamp
Material Malleable Cast Iron
Diameter (Metric) 15 mm
Length (Metric) 150 mm
Diameter (Metric) Inside 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.