missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydroxide, 95%, pure

Product Code. 10185520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185520 500 g Plastic bottle 500g
10521574 25 g Plastic bottle 25g
10572734 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10185520 Supplier Thermo Scientific Acros Supplier No. 315725000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 94% min. (Complexometry)
Linear Formula Mg(OH)2
Merck Index 15, 5734
Solubility Information Solubility in water: insoluble
IUPAC Name magnesium(2+) dihydroxide
Formula Weight 58.33
Percent Purity 95%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium hydroxide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium hydroxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.