missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAG Polyclonal Antibody
GREENER_CHOICE

Product Code. 10087562
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10087562 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10087562 Supplier Invitrogen™ Supplier No. 346200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

MAG Polyclonal Antibody for IHC (F), IP, Western Blot

The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAG
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene MAG
Gene Accession No. P07722, P20916, P20917
Gene Alias 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75
Gene Symbols MAG
Host Species Rabbit
Immunogen Human S/L-MAG fusion protein with a C-terminal region insert.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17136, 29409, 4099
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.