missing translation for 'onlineSavingsMsg'
Learn More

MAC13243, MedChemExpress

Product Code. 16478677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478677 5 mg 5mg
16446078 10 mg 10mg
16432217 50 mg 50mg
16426476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16478677 Supplier Medchem Express Supplier No. HY14456A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MAC13243, an antibacterial agent, is an inhibitor of bacterial lipoprotein targeting chaperone, LolA. MAC13243 is an antibacterial agent with Gram-negative selectivity.

MAC13243, an antibacterial agent, is an inhibitor of bacterial lipoprotein targeting chaperone, LolA. MAC13243 is an antibacterial agent with Gram-negative selectivity.

Specifications

Chemical Name or Material MAC13243
Physical Form Solid
CAS 1071638-38-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H25Cl2N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (113.02 mM; Need ultrasonic)
SMILES ClC1=CC=C(CSC2=NCN(CCC3=CC(OC)=C(OC)C=C3)CN2)C=C1.Cl
Molecular Weight (g/mol) 442.4
Formula Weight 442.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.