missing translation for 'onlineSavingsMsg'
Learn More

M2I-1, MedChemExpress

Product Code. 16470006
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470006 2 mg 2mg
16454276 100 mg 100mg
16439526 50 mg 50mg
16435697 5 mg 5mg
16432556 25 mg 25mg
16412958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16470006 Supplier Medchem Express Supplier No. HY100341

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress M2I-1 is a Mad2 inhibitor targeting the binding of Mad2 to Cdc20, an essential protein-protein interaction (PPI) within the spindle assembly checkpoint (SAC).

M2I-1 is a Mad2 inhibitor targeting the binding of Mad2 to Cdc20, an essential protein-protein interaction (PPI) within the spindle assembly checkpoint (SAC).

Specifications

Chemical Name or Material M2I-1
Physical Form Solid
CAS 312271-03-7
Color Orange
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C19H24N4O4S
Solubility Information DMSO : ≥ 32 mg/mL (79.11 mM)
SMILES O=C(/C(C(N1)=O)=C\C2=CC=C(N(CC(C)C)CC(C)C)C([N+]([O-])=O)=C2)NC1=S
Molecular Weight (g/mol) 404.48
Formula Weight 404.48
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.