missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M13 Phage coat protein Monoclonal Antibody (MM05)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for The antibody binds specifically to phage coat proteins of fd phage or M13 phage.
M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
Specifications
Specifications
| Antigen | M13 Phage coat protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MM05 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% Tween 80 and no preservative; pH 7.4 |
| Gene Alias | Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein |
| Host Species | Mouse |
| Immunogen | M13 Bacteriophage. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?