Learn More
Julabo™ M+R in-line Pt100 Sensor for Cryo-compact Circulator
Specifications
Specifications
| For Use With (Equipment) | Models FC1600T and FCW2500T |
| Type | Pt100 Sensor |
| For Use With (Application) | For measurement and control in external system |
| Includes | 2 Fittings M16 x 1 male and a 1.5m cable |