missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) nitrate hydrate, 99.9% (REO)

Product Code. p-7049508
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302518 1 g 1g
11312518 5 g 5g
11420470 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302518 Supplier Thermo Scientific Alfa Aesar Supplier No. 014568.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100641-16-5 | LuN3O9 | 360.98 g/mol

Lutetium(III) nitrate hydrate is used in ceramics, glass, phosphors and in lasers. It finds application as a catalyst in petroleum cracking and to prepare laser crystal. Further, it is used in hydrogenation, alkylation and polymerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100641-16-5
Molecular Formula LuN3O9
Molecular Weight (g/mol) 360.98
MDL Number MFCD00150421
InChI Key APRNQTOXCXOSHO-UHFFFAOYSA-N
Synonym lutetium iii nitrate hydrate, lutetium nitrate hydrate, acmc-1buji, lu.3no3.h2o, lutetium nitrate-water 1/3/1, lutetium 3+ hydrate trinitrate
PubChem CID 16212852
SMILES [Lu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 1 g
Linear Formula Lu(NO3)3·xH2O
UN Number UN1477
Solubility Information Soluble in water. Slightly soluble in strong mineral acids.
IUPAC Name lutetium(3+);trinitrate;hydrate
Formula Weight 360.98 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lutetium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.