Learn More
Thermo Scientific™ Oxoid™ Luria Broth (Miller's Modified)
Description
Miller's modified luria broth from Oxoid is used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Specifications
Specifications
| Product Type | Microbiology Media |
| For Use With (Application) | For the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins |
| Quantity | 500 g |