Learn More
Hamilton™ Male Luer Lock
Description
- Provided individually
- A range of thread sizes available
Specifications
Specifications
| Type | Male Luer Lock |
| Thread Size | 1/4 in. - 28 UNF Thread |
| For Use With | Thread depth less than 8 mm |