missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lucifer Yellow Polyclonal Antibody, Biotin

Description
Lucifer Yellow Polyclonal Antibody for IHC, ICC/IF
Even though the cell soma of a lucifer yellow-filled neuron may be brightly stained, its finer processes can sometimes be faint and may fade rapidly or be obscured by the more intensely stained portions of the neuron. By using anti-lucifer yellow dye antibodies in conjunction with standard enzyme-mediated immunohistochemical methods one can develop a more permanent, fade-free signal for light microscopy.
Specifications
Specifications
| Antigen | Lucifer Yellow |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 5mM sodium azide; pH 7.2 |
| Gene Alias | Lucifer Yellow Dye |
| Host Species | Rabbit |
| Immunogen | Lucifer Yellow. |
| Purification Method | IgG fraction |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?