missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Silica HR
Description
Includes
- Aqua Comparator Housing
- Aqua Comparator Disc
- Silica HR Acid Reagent (100 pack)
- Silica HR Citric Acid Reagent (100 pack)
- SilicaHR Molybdate Reagent (100 pack)
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Specifications
| Accuracy | ±5% |
| For Use With (Application) | Routine water analysis |
| Includes | Aqua comparator™ housing, aqua comparator™ disc, silica HR acid reagent, silica HR citric acid reagent, silica HR molybdate reagent, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
| No. of Tests | 100 |
| Range | 0 to 100mg/L |
| Type | HR Silica Test Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?