missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide Oxide, Mikromol™
Description
Loperamide Oxide
Specifications
Specifications
| Chemical Name or Material | Loperamide Oxide |
| CAS | 106900-12-3 |
| Analyte or Component Names | Loperamide Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H33 Cl N2 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Loperamide Hydrochloride Imp. F (EP), Loperamide Imp. F (EP), Loperamide USP Related Compound F, Loperamide USP RC F, Loperamide Oxide, trans-4-(4-chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutanamide 1-oxide, Arestal, Loperamide oxide, R 58425, Loperamide Related Compound F, Loperamide Hydrochloride Impurity F, Loperamide Impurity F |
| InChI Formula | InChI=1S/C29H33ClN2O3/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32(35)20-17-28(34,18-21-32)23-13-15-26(30)16-14-23/h3-16,34H,17-22H2,1-2H3/t28-,32- |
| SMILES | CN(C)C(=O)C(CC[N@+]1([O-])CC[C@](O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxy-1-oxidopiperidin-1-ium-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?