missing translation for 'onlineSavingsMsg'
Learn More

LKB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10009019
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10009019 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10009019 Supplier Invitrogen Supplier No. PA514072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LKB1 Polyclonal Antibody

LKB1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen LKB1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STK11
Gene Accession No. Q15831
Gene Alias AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11
Gene Symbols STK11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 14-44 amino acids from the N-terminal region of human STK11 (LKB1).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.