missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, 99%, extra pure

Product Code. 10032530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032530 250 g Plastic bottle 250g
10582981 25 g Plastic bottle 25g
10637694 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10032530 Supplier Thermo Scientific Acros Supplier No. 194722500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13453-78-6 | ClLiO4·3H2O | 160.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-78-6
Molecular Formula ClLiO4·3H2O
Molecular Weight (g/mol) 160.44
MDL Number MFCD00149765
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M
Synonym lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
PubChem CID 23708903
IUPAC Name lithium;perchlorate;trihydrate
SMILES [Li+].O.O.O.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 95°C
Color White
pH 6.0 to 7.5 (5% soln. at 20°C)
Physical Form Crystals and/or Chunks
Quantity 250 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiClO4·3H2O
Merck Index 15, 5595
Solubility Information Solubility in water: 597g/L water (25°C). Other solubilities: soluble in alcohol
Formula Weight 160.44
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium perchlorate trihydrate, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium perchlorate trihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.