missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium carbonate, 99.999%, (trace metal basis), extra pure
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Melting Point | 720.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.999% |
| Linear Formula | Li2CO3 |
| Fieser | 01,606; 03,183; 04,192; 05,396 |
| Merck Index | 15, 5583 |
| Solubility Information | Solubility in water: 13g/L (20°C). Other solubilities: soluble in diluted acids, practically insoluble in alcohol |
| IUPAC Name | dilithium(1+) carbonate |
| Show More |
Safety and Handling
Product Identifier
- Lithium carbonate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Lithium carbonate
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?