missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Liq-Glass

Product Code. 10129390
Change view
Click to view available options
Diameter (Metric):
12 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10129390 12 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10129390 Supplier Hamilton™ Supplier No. 238000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Connector Types S7
pH Range 0 to 14
Type Liq-Glass
For Use With (Application) For daily laboratory use
Length (Metric) 120 mm
Diameter (Metric) 12 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.