Learn More
JULABO™ Lift-up Makrolon™ Bath Cover for SW22/23
Description
This accessory is suitable for JULABO™ SW22, SW23
Specifications
Specifications
| For Use With (Equipment) | SW22, SW23, TW20 shaking water bath |
| Type | Bath Cover |
| For Use With (Application) | Prevent liquid losses due to evaporation, protects samples from contamination to 80°C |
| Material | Makrolon |