Learn More
BOLA Liebig Condensers Vacuum
Description
- One-piece cooling tube with ground joint socket and cone size 29 made of PTFE, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water.
- The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Includes | hose connectors made of PP and nuts for connection |
| Length (Metric) Jacket | 450 mm |