missing translation for 'onlineSavingsMsg'
Learn More

Levomefolic Acid, TRC

Product Code. 30008029
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008029 50 mg 50mg
30008215 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008029 Supplier Toronto Research Chemicals Supplier No. TRCL37573050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31690-09-2
Molecular Formula C20 H25 N7 O6
Molecular Weight (g/mol) 459.46
Synonym Glutamic acid, N-[p-[[(2-amino-5,6,7,8-tetrahydro-4-hydroxy-5-methyl-6-pteridinyl)methyl]amino]benzoyl]-, L-(-)- (8CI), L-Glutamic acid, N-[4-[[(2-amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-, (S)-, L-Glutamic acid, N-[4-[[[(6S)-2-amino-1,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl]methyl]amino]benzoyl]- (9CI), N-[4-[[[(6S)-2-Amino-3,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl]methyl]amino]benzoyl]-L-glutamic acid, (6S)-5-Methyltetrahydrofolic acid, 5-Methyl-(6S)-tetrahydrofolic acid, L-Methylfolate, LMSR, l-N5-Methyl-L-tetrahydrofolic acid, L-Glutamic acid, N-[4-[[[(6S)-2-amino-3,4,5,6,7,8-hexahydro-5-methyl-4-oxo-6-pteridinyl]methyl]amino]benzoyl]-, Levomefolic acid
IUPAC Name (2S)-2-[[4-[[(6S)-2-amino-5-methyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl]methylamino]benzoyl]amino]pentanedioic acid
SMILES CN1[C@@H](CNc2ccc(cc2)C(=O)N[C@@H](CCC(=O)O)C(=O)O)CNC3=C1C(=O)N=C(N)N3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H25N7O6/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,12-13,22H,6-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31)/t12-,13-/m0/s1
Formula Weight 459.1866
Chemical Name or Material Levomefolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.