missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levomefolic acid, MedChemExpress
Description
Levomefolic acid (5-MTHF) is an orally active, brain-penetrant natural active form of folic acid and is one of the most widely used folic acid food supplements.
Specifications
Specifications
| Chemical Name or Material | Levomefolic acid |
| Physical Form | Solid |
| CAS | 31690-09-2 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H25N7O6 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | 5-MTHF |
| Solubility Information | H2O : 5 mg/mL (10.88 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.18 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?