Learn More
Emma Safety Footwear Leo All-Round Safety XD-Fit Shoes
Product Code. 10612715
Change view
Click to view available options
Size:
41
42
43
44
45
46
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10612715 45 1 pair
10749425 43 1 pair
10431894 46 1 pair
10452694 42 1 pair
10295384 44 1 pair
10194154 41 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10612715 Supplier Emma Safety Footwear Supplier No. MM70156845
Only null left
Add to basket
Add to basket
Designed for people with slightly broader-than-normal feet.
  • Full grain leather uppers
  • Hydro-Tec™ sanitized silver lining
  • Non-woven antistatic insole
  • GripForce™ Easy Twist outsole
  • Strobel-stitched upper construction
Specifications
Color Black
Liner Material Hydro-Tec Sanitized Silver lining
Material Leather
Size 45
Sole Features Non-woven antistatic insole
Sole Material GripForce™ Easy Twist outsole
Toe Material Steel
Product Type All-Round Safety Shoes